Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
LSM12
Entrez Gene ID
856521
Gene Name
Gene Aliases
YHR121W
Chromosome Location
Chr.VIII: 352756 - 353319 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179251.1 NP_011989.1 1 - 1 452 99

View Full Product Details