Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCY1
Entrez Gene ID
854778
Gene Name
Gene Aliases
SRA1, YIL033C
Chromosome Location
Chr.IX: 290419 - 291669 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179383.1 NP_012231.1 1 - 1 1149 73

View Full Product Details