Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CKA1
Entrez Gene ID
854776
Gene Name
Gene Aliases
YIL035C
Chromosome Location
Chr.IX: 287790 - 288908 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179385.1 NP_012229.1 1 - 1 950 88

View Full Product Details