Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RRT14
Entrez Gene ID
854679
Gene Name
Gene Aliases
YIL127C
Chromosome Location
Chr.IX: 117024 - 117644 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179475.1 NP_012139.1 1 - 1 452 91

View Full Product Details