Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBP7
Entrez Gene ID
854650
Gene Name
Gene Aliases
YIL156W
Chromosome Location
Chr.IX: 48091 - 51306 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179504.1 NP_012110.1 1 - 1 3047 95

View Full Product Details