Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DAL3
Entrez Gene ID
854850
Gene Name
Gene Aliases
YIR032C
Chromosome Location
Chr.IX: 415030 - 415617 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179554.1 NP_012298.1 1 - 1 448 70

View Full Product Details