Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
GWT1
Entrez Gene ID
853354
Gene Name
Gene Aliases
YJL091C
Chromosome Location
Chr.X: 261080 - 262552 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181524.1 NP_012444.2 1 - 1 1352 93

View Full Product Details