Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SET4
Entrez Gene ID
853339
Gene Name
Gene Aliases
YJL105W
Chromosome Location
Chr.X: 225055 - 226737 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181538.1 NP_012430.1 1 - 1 1548 94

View Full Product Details