Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AVT1
Entrez Gene ID
853457
Gene Name
Gene Aliases
YJR001W
Chromosome Location
Chr.X: 436802 - 438610 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181658.1 NP_012534.1 1 - 1 1449 89

View Full Product Details