Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SSC1
Entrez Gene ID
853503
Gene Name
Gene Aliases
ENS1, YJR045C
Chromosome Location
Chr.X: 519638 - 521602 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181703.1 NP_012579.1 1 - 1 1857 93

View Full Product Details