Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RPL43B
Entrez Gene ID
853557
Gene Name
Gene Aliases
YJR094W-A
Chromosome Location
Chr.X: 608305 - 608858 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181752.3 NP_012628.3 1 - 1 48 104

View Full Product Details