Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DEF1
Entrez Gene ID
853811
Gene Name
Gene Aliases
VID31, YKL054C
Chromosome Location
Chr.XI: 336537 - 338753 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179620.1 NP_012869.1 1 - 1 1954 71

View Full Product Details