Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YPK1
Entrez Gene ID
853733
Gene Name
Gene Aliases
SLI2, YKL126W
Chromosome Location
Chr.XI: 205707 - 207749 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179692.1 NP_012796.1 1 - 1 1751 98

View Full Product Details