Assay Design

The amplified product spans an exon junction and the probe and one of the primers sit within one exon. This assay may detect gDNA.

Gene Transcripts

Gene Symbol
OCT1
Entrez Gene ID
853724
Gene Name
Gene Aliases
YKL134C
Chromosome Location
Chr.XI: 189124 - 191442 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179700.2 NP_012788.2 1 - 1 2260 91

View Full Product Details