Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HSK3
Entrez Gene ID
853719
Gene Name
Gene Aliases
YKL138C-A
Chromosome Location
Chr.XI: 184803 - 185012 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184513.1 NP_690844.1 1 - 1 145 95

View Full Product Details