Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZRT3
Entrez Gene ID
853679
Gene Name
Gene Aliases
YKL175W
Chromosome Location
Chr.XI: 118793 - 120304 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179741.1 NP_012746.1 1 - 1 1448 92

View Full Product Details