Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BCH2
Entrez Gene ID
853898
Gene Name
Gene Aliases
FMP50, YKR027W
Chromosome Location
Chr.XI: 491364 - 493661 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179817.3 NP_012952.3 1 - 1 1849 83

View Full Product Details