Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CAF4
Entrez Gene ID
853908
Gene Name
Gene Aliases
YKR036C
Chromosome Location
Chr.XI: 508702 - 510633 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001179826.2 NP_012962.3 1 - 1 1850 91

View Full Product Details