Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HIF1
Entrez Gene ID
850638
Gene Name
Gene Aliases
YLL022C
Chromosome Location
Chr.XII: 99044 - 100201 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181842.1 NP_013078.1 1 - 1 951 103

View Full Product Details