Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FPS1
Entrez Gene ID
850683
Gene Name
Gene Aliases
YLL043W
Chromosome Location
Chr.XII: 49938 - 51947 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001181863.1 NP_013057.1 1 - 1 1950 82

View Full Product Details