Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACS2
Entrez Gene ID
850846
Gene Name
Gene Aliases
YLR153C
Chromosome Location
Chr.XII: 445524 - 447575 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182040.1 NP_013254.1 1 - 1 1952 101

View Full Product Details