Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MDL1
Entrez Gene ID
850885
Gene Name
Gene Aliases
YLR188W
Chromosome Location
Chr.XII: 528300 - 530387 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182075.1 NP_013289.1 1 - 1 1946 84

View Full Product Details