Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEC61
Entrez Gene ID
851095
Gene Name
Gene Aliases
YLR378C
Chromosome Location
Chr.XII: 875736 - 877178 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182267.1 NP_013482.1 1 - 1 1255 86

View Full Product Details