Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ATP10
Entrez Gene ID
851109
Gene Name
Gene Aliases
YLR393W
Chromosome Location
Chr.XII: 907079 - 907918 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182281.1 NP_013497.1 1 - 1 749 97

View Full Product Details