Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPC2
Entrez Gene ID
854949
Gene Name
Gene Aliases
SPY1, YML055W
Chromosome Location
Chr.XIII: 164790 - 165326 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182413.1 NP_013657.1 1 - 1 347 89

View Full Product Details