Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
NTE1
Entrez Gene ID
854943
Gene Name
Gene Aliases
YML059C
Chromosome Location
Chr.XIII: 153219 - 158258 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182418.1 NP_013652.1 1 - 1 4952 98

View Full Product Details