Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PLB2
Entrez Gene ID
855018
Gene Name
Gene Aliases
YMR006C
Chromosome Location
Chr.XIII: 277561 - 279681 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182502.1 NP_013719.1 1 - 1 2048 93

View Full Product Details