Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
UBX4
Entrez Gene ID
855089
Gene Name
Gene Aliases
CUI1, YMR067C
Chromosome Location
Chr.XIII: 404323 - 405573 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182565.1 NP_013783.1 1 - 1 1147 65

View Full Product Details