Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAA4
Entrez Gene ID
855288
Gene Name
Gene Aliases
YMR246W
Chromosome Location
Chr.XIII: 759807 - 761891 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182754.1 NP_013974.1 1 - 1 1846 89

View Full Product Details