Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
AEP2
Entrez Gene ID
855325
Gene Name
Gene Aliases
ATP13, YMR282C
Chromosome Location
Chr.XIII: 833356 - 835098 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182789.1 NP_014009.1 1 - 1 1350 84

View Full Product Details