Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SIW14
Entrez Gene ID
855699
Gene Name
Gene Aliases
OCA3, YNL032W
Chromosome Location
Chr.XIV: 574506 - 575351 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182871.3 NP_014366.3 1 - 1 751 95

View Full Product Details