Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TCB2
Entrez Gene ID
855637
Gene Name
Gene Aliases
YNL087W
Chromosome Location
Chr.XIV: 462411 - 465947 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182925.1 NP_014312.1 1 - 1 3448 89

View Full Product Details