Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAR11
Entrez Gene ID
855596
Gene Name
Gene Aliases
YNL127W
Chromosome Location
Chr.XIV: 383987 - 386848 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001182965.3 NP_014272.3 1 - 1 2752 99

View Full Product Details