Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FMP41
Entrez Gene ID
855553
Gene Name
Gene Aliases
YNL168C
Chromosome Location
Chr.XIV: 318030 - 318809 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183006.1 NP_014231.1 1 - 1 642 72

View Full Product Details