Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
RIO2
Entrez Gene ID
855514
Gene Name
Gene Aliases
YNL207W
Chromosome Location
Chr.XIV: 255353 - 256630 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183045.1 NP_014192.1 1 - 1 1149 80

View Full Product Details