Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PPG1
Entrez Gene ID
855766
Gene Name
Gene Aliases
YNR032W
Chromosome Location
Chr.XIV: 686010 - 687116 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183209.3 NP_014429.3 1 - 1 845 96

View Full Product Details