Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CMK2
Entrez Gene ID
854144
Gene Name
Gene Aliases
YOL016C
Chromosome Location
Chr.XV: 294777 - 296120 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183270.1 NP_014626.1 1 - 1 1250 104

View Full Product Details