Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CUE5
Entrez Gene ID
854206
Gene Name
Gene Aliases
YOR042W
Chromosome Location
Chr.XV: 408425 - 409660 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183461.1 NP_014685.1 1 - 1 946 82

View Full Product Details