Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MSA1
Entrez Gene ID
854232
Gene Name
Gene Aliases
YOR066W, YOR29-17
Chromosome Location
Chr.XV: 449436 - 451325 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183485.1 NP_014709.1 1 - 1 1653 78

View Full Product Details