Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
OST3
Entrez Gene ID
854252
Gene Name
Gene Aliases
YOR085W
Chromosome Location
Chr.XV: 482033 - 483085 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183504.1 NP_014728.1 1 - 1 948 88

View Full Product Details