Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
YVC1
Entrez Gene ID
854255
Gene Name
Gene Aliases
YOR087W, YOR088W
Chromosome Location
Chr.XV: 487707 - 489734 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183506.1 NP_014730.2 1 - 1 1952 83

View Full Product Details