Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ECM3
Entrez Gene ID
854259
Gene Name
Gene Aliases
YOR092W, YOR3165W
Chromosome Location
Chr.XV: 495127 - 496968 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183511.1 NP_014735.1 1 - 1 1550 66

View Full Product Details