Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
BFR1
Entrez Gene ID
854373
Gene Name
Gene Aliases
YOR198C
Chromosome Location
Chr.XV: 718653 - 720065 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183617.1 NP_014841.1 1 - 1 1254 94

View Full Product Details