Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WTM2
Entrez Gene ID
854404
Gene Name
Gene Aliases
YOR229W
Chromosome Location
Chr.XV: 768409 - 769812 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183648.1 NP_014872.1 1 - 1 1351 69

View Full Product Details