Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SRL1
Entrez Gene ID
854421
Gene Name
Gene Aliases
YOR247W
Chromosome Location
Chr.XV: 797676 - 798308 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183666.1 NP_014890.1 1 - 1 451 93

View Full Product Details