Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MCH5
Entrez Gene ID
854483
Gene Name
Gene Aliases
YOR306C
Chromosome Location
Chr.XV: 889867 - 891432 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183726.4 NP_014951.4 1 - 1 1356 79

View Full Product Details