Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SPS4
Entrez Gene ID
854490
Gene Name
Gene Aliases
YOR313C
Chromosome Location
Chr.XV: 901858 - 902874 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183733.3 NP_014958.3 1 - 1 951 103

View Full Product Details