Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FAA1
Entrez Gene ID
854495
Gene Name
Gene Aliases
YOR317W
Chromosome Location
Chr.XV: 909343 - 911445 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183737.3 NP_014962.3 1 - 1 2050 79

View Full Product Details