Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
TEA1
Entrez Gene ID
854515
Gene Name
Gene Aliases
YOR337W
Chromosome Location
Chr.XV: 954344 - 956623 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183757.3 NP_014982.3 1 - 1 2148 83

View Full Product Details