Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PDR12
Entrez Gene ID
856049
Gene Name
Gene Aliases
YPL058C
Chromosome Location
Chr.XVI: 445842 - 450377 on Build R64
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001183872.1 NP_015267.1 1 - 1 4456 100

View Full Product Details